Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CIRH1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109545
Description
Immunogen sequence: NWLAASGTSA GVHVYNVKQL KLHCTVPAYN FPVTAMAIAP NTNNLVIAHS DQQVFEYSIP DKQYTDWSRT VQKQGFHHLW LQRDTPITHI SFHPKRPMHI LLHDAYMFCI IDKSLPLPND KTLLYNPFPP TNESDVIRRR TAHAFKISKI YKPLLFMDLL DERTLVAVER PLDDIIAQLP PPIKKKKFGT.
CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
Specifications
| CIRH1A | |
| Polyclonal | |
| Unconjugated | |
| UTP4 | |
| CIRH1A; Cirhin; cirrhosis, autosomal recessive 1A (cirhin); cirrhosis, autosomal recessive 1A (human); cPERP-E; Kiaa1988; Naic; Teg-292; testis expressed gene 292; testis-expressed gene 292; testis-expressed gene 292 protein; Tex292; U3 small nucleolar RNA-associated protein 4 homolog; UTP4; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog; UTP4, small subunit processome component | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21771, 84916 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R2N2, Q969X6 | |
| UTP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 497-686 of human CIRH1A (NP_116219.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction