Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIRH1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111179 Supplier Invitrogen™ Supplier No. PA5111179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVRFFNYVPS GIRCVAYNNQ SNRLAVSRTD GTVEIYNLSA NYFQEKFFPG HESRATEALC WAEGQRLFSA GLNGEIMEYD LQ Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 96%.

CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIRH1A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP4
Gene Accession No. Q969X6
Gene Alias CIRH1A; Cirhin; cirrhosis, autosomal recessive 1A (cirhin); cirrhosis, autosomal recessive 1A (human); cPERP-E; Kiaa1988; Naic; Teg-292; testis expressed gene 292; testis-expressed gene 292; testis-expressed gene 292 protein; Tex292; U3 small nucleolar RNA-associated protein 4 homolog; UTP4; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog; UTP4, small subunit processome component
Gene Symbols UTP4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CIRH1A. Recombinant protein control fragment (Product #RP-106044).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.