Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™

Catalog No. AC138230010
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle

Chemical Identifiers

CAS 1469-48-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00005880
InChI Key CIFFBTOJCKSRJY-OLQVQODUSA-N
Synonym cis-1,2,3,6-tetrahydrophthalimide, cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione, cis-4-cyclohexene-1,2-dicarboximide, cis-tetrahydrophthalimide, unii-es57d28o1u, 4-cyclohexene-1,2-dicarboximide, cis, cis-.delta.4-tetrahydrophthalimide, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione
PubChem CID 92888
IUPAC Name (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
SMILES C1C=CCC2C1C(=O)NC2=O

Specifications

Melting Point 131.0°C to 138.0°C
Color Beige to Brown-Yellow or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Quantity 1 kg
Formula Weight 151.16
Percent Purity 96%
Physical Form Powder or Flakes
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalimide, 96%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-999-
RTECSNumber GW4635000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.