Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2,3,6-Tetrahydrophthalimide, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
250g
1kg
Packaging:
Glass bottle
Plastic bottle
1469-48-3
C8H9NO2
151.16
MFCD00005880
CIFFBTOJCKSRJY-OLQVQODUSA-N
cis-1,2,3,6-tetrahydrophthalimide,cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione,cis-4-cyclohexene-1,2-dicarboximide,cis-tetrahydrophthalimide,unii-es57d28o1u,4-cyclohexene-1,2-dicarboximide, cis,cis-.delta.4-tetrahydrophthalimide,1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis,1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel,3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione
92888
(3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
C1C=CCC2C1C(=O)NC2=O
1469-48-3
C8H9NO2
151.16
MFCD00005880
CIFFBTOJCKSRJY-OLQVQODUSA-N
cis-1,2,3,6-tetrahydrophthalimide,cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione,cis-4-cyclohexene-1,2-dicarboximide,cis-tetrahydrophthalimide,unii-es57d28o1u,4-cyclohexene-1,2-dicarboximide, cis,cis-.delta.4-tetrahydrophthalimide,1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis,1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel,3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione
92888
(3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
C1C=CCC2C1C(=O)NC2=O
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige to Brown-Yellow or Yellow
Melting Point 131.0°C to 138.0°C
Assay Percent Range 96%
Formula Weight 151.16
Physical Form Powder or Flakes
Percent Purity 96%
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalimide, 96%

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-999-

RTECSNumber : GW4635000