Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Specifications
Specifications
Melting Point | 131.0°C to 138.0°C |
Color | Beige to Brown-Yellow or Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% |
Packaging | Plastic bottle |
Quantity | 250 g |
Formula Weight | 151.16 |
Percent Purity | 96% |
Physical Form | Powder or Flakes |
Chemical Name or Material | cis-1,2,3,6-Tetrahydrophthalimide, 96% |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement: Harmful if inhaled. Harmful in contact with skin. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 215-999- |
RTECSNumber | GW4635000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction