Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific™

Catalog No. AC299200250
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-09-3
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00064949
InChI Key QSAWQNUELGIYBC-OLQVQODUSA-L
Synonym cis-1,2-cyclohexanedicarboxylic acid, cis-cyclohexane-1,2-dicarboxylic acid, cis-hexahydrophthalic acid, 1r,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2r-cyclohexane-1,2-dicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2s-rel, cis-1,2-cyclohexanedicarboxylicacid, ksc497o9d, cis-+/--perhydrophthalic acid, cis-cyclohexane-1,2-dicarboxylicacid
PubChem CID 245564
IUPAC Name (1R,2S)-cyclohexane-1,2-dicarboxylate
SMILES [O-]C(=O)[C@H]1CCCC[C@H]1C([O-])=O

Specifications

Melting Point 188°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Formula Weight 172.18
Percent Purity ≥97.5%
Physical Form Crystals
Chemical Name or Material cis-1, 2-Cyclohexanedicarboxylic acid, 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.