Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Cyclohexanediol, 99%

Catalog No. p-3757350
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 1792-81-0 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1792-81-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00064944
InChI Key PFURGBBHAOXLIO-OLQVQODUSA-N
Synonym cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
PubChem CID 92903
ChEBI CHEBI:32329
IUPAC Name (1R,2S)-cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 98°C to 101°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 1 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 116.16
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material cis-1, 2-Cyclohexanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.