Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Dichloro-2-butene, 95%

Catalog No. AC187601000
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1476-11-5 | C4H6Cl2 | 125 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-11-5
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 125
InChI Key FQDIANVAWVHZIR-UPHRSURJSA-N
Synonym cis-1,4-dichloro-2-butene, z-1,4-dichlorobut-2-ene, 1,4-dichloro-cis-2-butene, 2-butene, 1,4-dichloro-, z, cis-1,4-dichlorobut-2-ene, unii-bqs2752i90, ccris 2651, cis 1,4-dichlorobutene, 2-butene, 1,4-dichloro-, cis, cis-2,3-dichlorobut-2-ene
PubChem CID 6432115
IUPAC Name (Z)-1,4-dichlorobut-2-ene
SMILES C(C=CCCl)Cl

Specifications

Melting Point -48.0°C
Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 152.0°C (758.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4884 to 1.4914
Linear Formula ClCH2CH=CHCH2Cl
Quantity 100 mL
Beilstein ,III,743
Fieser 10,134; 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270
Specific Gravity 1.18
Formula Weight 125
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-1, 4-Dichloro-2-butene
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 216-021-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.