Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-11-Hexadecenal 95.0+%, TCI America™
SDP

Catalog No. h06791g
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 53939-28-9
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.42
MDL Number MFCD00010124
InChI Key AMTITFMUKRZZEE-WAYWQWQTSA-N
Synonym cis-11-Hexadecenyl Aldehyde
PubChem CID 5364495
IUPAC Name (11Z)-hexadec-11-enal
SMILES CCCC\C=C/CCCCCCCCCC=O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 1 g
Formula Weight 238.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-11-Hexadecenal
EINECSNumber (2)-3385
RTECSNumber MM0350000
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.