Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Aminocyclohexanol hydrochloride, 95%

Catalog No. p-3735287
Click to view available options
Quantity:
1 g
2.5 g
5 g

CAS: 6936-47-6 | C6H13NO·HCl | 151.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6936-47-6
MDL Number MFCD00143981
InChI Key LKKCSUHCVGCGFA-RIHPBJNCSA-N
Synonym 1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
PubChem CID 12228413
IUPAC Name (1R,2S)-2-aminocyclohexan-1-ol;hydrochloride
SMILES C1CCC(C(C1)N)O.Cl

Specifications

Melting Point 186.0°C to 190.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H13NO·HCl
Linear Formula H2NC6H10OH·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,hot ethanol
Molecular Weight (g/mol) 151.64
Formula Weight 151.64
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Aminocyclohexanol hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.