Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer

Catalog No. p-7021785
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1576-95-0 | C5H10O | 86.13 g/mol

cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-95-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00063208
InChI Key BTSIZIIPFNVMHF-ARJAWSKDSA-N
Synonym cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
PubChem CID 5364919
SMILES CC\C=C/CO

Specifications

Density 0.853
Boiling Point 138°C
Flash Point 48°C (118°F)
Refractive Index 1.436
Quantity 5 g
UN Number UN1987
Beilstein 1719473
Solubility Information Miscible with alcohol. Immiscible with water.
IUPAC Name (2Z)-pent-2-en-1-ol
Formula Weight 86.13
Percent Purity 97%
Assay remainder mainly trans-isomer
Chemical Name or Material cis-2-Penten-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 216-415-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.