Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Pentene

Catalog No. p-3523402
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle

CAS: 627-20-3 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-20-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00063978
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z
PubChem CID 5326160
IUPAC Name (Z)-pent-2-ene
SMILES CC\C=C/C

Specifications

Melting Point -151.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 37.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3810 to 1.3830 (20°C, 589nm)
Linear Formula CH3CHCHCH2CH3
Quantity 1 g
Specific Gravity 0.65
Merck Index 15, 7235
Solubility Information Solubility in water: insoluble.
Formula Weight 70.13
Percent Purity ≥95.0%
Physical Form Liquid
Chemical Name or Material cis-2-Pentene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-988-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.