Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid, 97%, Thermo Scientific™

Catalog No. AAH6247503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6247503 1 g
AAH62475MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6247503 Supplier Thermo Scientific Chemicals Supplier No. H6247503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64396-97-0
Molecular Formula C10H16O3
Molecular Weight (g/mol) 184.24
MDL Number MFCD00001327,MFCD09864988
InChI Key SIZDUQQDBXJXLQ-UHFFFAOYNA-N
Synonym cis-dl-pinonic acid, 1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, pinonic acid, cis dl, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl-acetic acid, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid, 1s,3s-2,2-dimethyl-3-acetylcyclobutyl acetic acid, 1s,3s-+-cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid
PubChem CID 239867
IUPAC Name 2-[(1S,3S)-3-acetyl-2,2-dimethylcyclobutyl]acetic acid
SMILES CC(=O)C1CC(CC(O)=O)C1(C)C

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Formula Weight 184.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.