Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-(Boc-amino)cyclohexanemethanol, 97%

Catalog No. AAH6274403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6274403 1 g
AAH62744MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6274403 Supplier Thermo Scientific Chemicals Supplier No. H6274403
Add to Cart

CAS: 920966-16-1 | C12H23NO3 | 229.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920966-16-1
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844608
InChI Key LBJSEPNOVVUVJA-ZJUUUORDSA-N
Synonym tert-Butyl cis-(3-hydroxymethyl)cyclohexylcarbamate; cis-1-(Boc-amino)-3-(hydroxymethyl)cyclohexane
PubChem CID 83823767
IUPAC Name tert-butyl N-[(1S,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CO

Specifications

Quantity 1 g
Formula Weight 229.31
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclohexanemethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.