Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-ol, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
928-96-1
C6H12O
100.16
MFCD00063217
UFLHIIWVXFIJGU-ARJAWSKDSA-N
z-hex-3-en-1-ol,cis-3-hexen-1-ol,cis-3-hexenol,leaf alcohol,blatteralkohol,z-3-hexenol,z-3-hexen-1-ol,3-hexenol,3z-hex-3-en-1-ol,3-hexen-1-ol, z
5281167
CHEBI:28857
(Z)-hex-3-en-1-ol
CCC=CCCO
928-96-1
C6H12O
100.16
MFCD00063217
UFLHIIWVXFIJGU-ARJAWSKDSA-N
z-hex-3-en-1-ol,cis-3-hexen-1-ol,cis-3-hexenol,leaf alcohol,blatteralkohol,z-3-hexenol,z-3-hexen-1-ol,3-hexenol,3z-hex-3-en-1-ol,3-hexen-1-ol, z
5281167
CHEBI:28857
(Z)-hex-3-en-1-ol
CCC=CCCO
Packaging Glass bottle
Refractive Index 1.4380 to 1.4410
Boiling Point 156.0°C to 157.0°C
Quantity 50mL
Flash Point 59°C
Infrared Spectrum Authentic
Specific Gravity 0.848
Color Colorless to Yellow
Density 0.8480g/mL
Assay Percent Range 98%
Linear Formula C2H5CH=CHCH2CH2OH
Beilstein 01, I, 229
Merck Index 10, 4592
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and propylene glycol, miscible with oils
Formula Weight 100.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material cis-3-Hexen-1-ol
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning