Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

Catalog No. AAH5273106
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 171110-72-8 | C11H21ClN2O4 | 280.75 g/mol

cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171110-72-8
Molecular Formula C11H21ClN2O4
Molecular Weight (g/mol) 280.75
MDL Number MFCD03787927
InChI Key KFYCQKLSGMAVQH-UHFFFAOYNA-N
Synonym methyl 2s,4s-1-boc-4-aminopyrrolidine-2-carboxylate hydrochloride, 2s,4s-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-cis-4-amino-l-proline methyl ester hydrochloride, 2s,4s-1-boc-4-amino pyrrolidine-2-carboxylic acid methylester-hcl, 1-tert-butyl 2-methyl 2s,4s-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-cis-4-amino-l-proline methyl ester hydrochloride salt, 1-tert-butyl 2-methyl cis-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, 2s,4s-1-boc-4-amino pyrrolidine-2-carboxylic acid methylester hydrochloride, 4-aminopyrrolidine-1,2-dicarboxylatehydrochloride, s,s-1-boc-4-aminoproline methyl ester hydrochloride
PubChem CID 17998957
SMILES Cl.COC(=O)C1CC(N)CN1C(=O)OC(C)(C)C

Specifications

Melting Point ∽185°C (decomposition)
Quantity 5 g
Optical Rotation −40° (c=2 in Methanol)
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate;hydrochloride
Formula Weight 280.75
Percent Purity 97%
Chemical Name or Material cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.