Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%

Catalog No. AAH5344606
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 2746-19-2 | C9H8O3 | 164.16 g/mol

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-19-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00630580
InChI Key KNDQHSIWLOJIGP-RNGGSSJXSA-N
Synonym cis-5-norbornene-exo-2,3-dicarboxylic anhydride, 3ar,4r,7s,7as-rel-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, himic anhydride, unii-n1kqo6559b, norborn-5-ene-2exo,3exo-dicarboxylic acid anhydride, 4,7-methanoisobenzofuran-1,3-dione, 3a,4,7,7a-tetrahydro-, 3aalpha,4beta,7beta,7aalpha-9ci, exo-5-norbornene-2,3-dicarboxylic anhydride, exo-3,6-methylene-1,2,3,6-tetrahydrophthalic anhydride, endicanhydride, 1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
PubChem CID 637794
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 140°C to 145°C
Quantity 5 g
Formula Weight 164.16
Percent Purity 95%
Chemical Name or Material cis-5-Norbornene-exo-2,3-dicarboxylic anhydride
Hazard Category H317-H318-H334-H335
Hazard Statement GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
EINECSNumber 220-384-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.