Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals

Catalog No. p-3770887
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-19-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00630580
InChI Key KNDQHSIWLOJIGP-RNGGSSJXSA-N
Synonym cis-5-norbornene-exo-2,3-dicarboxylic anhydride, 3ar,4r,7s,7as-rel-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, himic anhydride, unii-n1kqo6559b, norborn-5-ene-2exo,3exo-dicarboxylic acid anhydride, 4,7-methanoisobenzofuran-1,3-dione, 3a,4,7,7a-tetrahydro-, 3aalpha,4beta,7beta,7aalpha-9ci, exo-5-norbornene-2,3-dicarboxylic anhydride, exo-3,6-methylene-1,2,3,6-tetrahydrophthalic anhydride, endicanhydride, 1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
PubChem CID 637794
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 144°C
Quantity 1 g
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Formula Weight 164.16
Percent Purity 95%
Chemical Name or Material cis-5-Norbornene-exo-2, 3-dicarboxylic anhydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-384-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.