Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Aconitic acid, tech. 85%

Catalog No. p-7022994
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 585-84-2 | C6H6O6 | 174.11 g/mol

cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

Solubility
Soluble in water, alcohol. Slightly soluble in ether.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-84-2
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.11
MDL Number MFCD00063184
InChI Key GTZCVFVGUGFEME-HNQUOIGGSA-N
Synonym cis-aconitic acid, cis-aconitate, achilleic acid, citridinic acid, equisetic acid, pyrocitric acid, achilleaic acid, citridic acid, cis-aconic acid, aconitic acid
PubChem CID 643757
ChEBI CHEBI:32805
SMILES OC(=O)C\C(=C/C(O)=O)C(O)=O

Specifications

Melting Point 115°C to 120°C
Assay Percent Range tech. 90%
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol. Slightly soluble in ether.
IUPAC Name (Z)-prop-1-ene-1,2,3-tricarboxylic acid
Formula Weight 174.11
Percent Purity 90%
Grade Technical
Chemical Name or Material cis-Aconitic Acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 209-561-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.