Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Diamminedichloroplatinum(II), Pt 64.5% min

Catalog No. AA1047103
Click to view available options
Quantity:
0.25 g
1 g
5 g

Potent Anticancer Agent That Blocks DNA Synthesis. | CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol

Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym Cisplatin
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl

Specifications

Melting Point 270°C (decomposition)
Color Yellow
Physical Form Micro-crystals
Quantity 1 g
Solubility Information Soluble in DMF. Insoluble in most common solvents
Formula Weight 300.06
Odor Odorless
Appearance Yellow micro crystals
Chemical Name or Material cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis
Hazard Category H300-H312+H332-H315-H318-H335-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packing Group: II
EINECSNumber 239-733-8
RTECSNumber TP2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.