Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)

Catalog No. AC193762500
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym cis-DDP
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl

Specifications

Melting Point 277.0°C
Color White
Physical Form Powder
Quantity 250 mg
Assay Percent Range 64.0 to 66% (Pt)
Linear Formula Pt(NH3)2Cl2
Merck Index 15, 2316
Solubility Information Solubility in water: 0.25g/100mL (25°C)
Formula Weight 300.05
Percent Purity 99.99%
Grade Trace Metal Basis
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material cis-Dichlorodiamineplatinum(II), 99.99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Fatal if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If
EINECSNumber 239-733-8
RTECSNumber TP2450000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.