Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis

Specifications
Specifications
Melting Point | 277.0°C |
Color | White |
Physical Form | Powder |
Quantity | 250 mg |
Assay Percent Range | 64.0 to 66% (Pt) |
Linear Formula | Pt(NH3)2Cl2 |
Merck Index | 15, 2316 |
Solubility Information | Solubility in water: 0.25g/100mL (25°C) |
Formula Weight | 300.05 |
Percent Purity | 99.99% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause cancer. Fatal if swallowed. Causes serious eye damage. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. If |
EINECSNumber | 239-733-8 |
RTECSNumber | TP2450000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction