Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Jasmone 92.0+%, TCI America™
SDP

Catalog No. J000325ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 488-10-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00001402,MFCD00001402,MFCD00001402
InChI Key XMLSXPIVAXONDL-PLNGDYQASA-N
Synonym trans-jasmone, e-jasmone, unii-7tcs3y45dr, 7tcs3y45dr, 3-methyl-2-e-pent-2-enyl cyclopent-2-en-1-one, 2-cyclopenten-1-one, 3-methyl-2-2-pentenyl-, e, 3-methyl-2-pent-2-en-1-yl cyclopent-2-en-1-one, 3-methyl-2-2e-pent-2-en-1-yl cyclopent-2-en-1-one, jasmone, e, 3-methyl-2-2-pentenyl-2-cyclopenten-1-one
PubChem CID 1549019
IUPAC Name 3-methyl-2-[(2Z)-pent-2-en-1-yl]cyclopent-2-en-1-one
SMILES CC\C=C/CC1=C(C)CCC1=O

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 mL
Formula Weight 164.25
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-Jasmone
EINECSNumber (3)-2394
RTECSNumber GY7301000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.