Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569873 Supplier Invitrogen™ Supplier No. PA569873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

CIS contains a SH2 domain and a SOCS box domain. The protein thus belongs to the cytokine-induced STAT inhibitor (CIS), also known as suppressor of cytokine signaling (SOCS) or STAT-induced STAT inhibitor (SSI), protein family. CIS family members are known to be cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by IL2, IL3, GM-CSF and EPO in hematopoietic cells. Proteasome-mediated degradation of this protein has been shown to be involved in the inactivation of the erythropoietin receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CISH
Gene Accession No. Q62225
Gene Alias AI385595; BACTS2; Cis; CIS1; CIS-1; Cish; CISH_tv2; cytokine inducible SH2 containing protein; cytokine inducible SH2 protein; cytokine inducible SH2-containing protein; cytokine inducible SH2-containing protein 1; cytokine-inducible inhibitor of signaling type 1B; cytokine-inducible SH2 protein; cytokine-inducible SH2-containing protein; F17; F23; G18; Protein G18; similar to Cytokine-inducible SH2-containing protein (CIS) (CIS-1) (Suppressor of cytokine signaling) (SOCS) (G18); SOCS; Suppressor of cytokine signaling
Gene Symbols CISH
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse CIS (aa 78-127).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12700
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.