Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-Stilbene, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
Solubility
Immiscible with water. Miscible with ethanol.
Notes
Incompatible with oxidizing agents.
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
Solubility
Immiscible with water. Miscible with ethanol.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 1°C to 2°C |
| Density | 1.04 |
| Boiling Point | 307°C |
| Flash Point | >110°C (230°F) |
| Odor | Weak |
| Refractive Index | 1.622 |
| Quantity | 5 g |
| Beilstein | 1616739 |
| Sensitivity | Light sensitive |
| Merck Index | 14,8817 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
| EINECSNumber | 211-445-7 |
| RTECSNumber | DA0513600 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction