Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ cis-VZ 185
GREENER_CHOICESDP

Catalog No. 69395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-395 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 69-395 Supplier Tocris Bioscience™ Supplier No. 6939/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Negative control for VZ 185 (Cat. No. 6936)

cis-VZ 185 is a negative control for VZ 185 (Cat. No. 6936).

Specifications

Chemical Name or Material (2 S,4 S)-N-(2-((5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyl)oxy)-4-(4-methylthiazol-5-yl)benzyl)-1-(-2-(1-fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide
CAS 2306193-98-4
Quantity 5 mg
Target Active Degrader Negative Controls
Molecular Formula C53H67FN8O8S
Formula Weight 2576
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.