Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Zeatin, MP Biomedicals™

Catalog No. ICN15227105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15227105 5 mg
ICN15227101 1 mg
ICN15227110 10 mg
ICN15227150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15227105 Supplier MP Biomedicals, Inc Supplier No. 0215227105

Chemical Identifiers

CAS 32771-64-5
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.248
MDL Number MFCD00070217
InChI Key UZKQTCBAMSWPJD-UQCOIBPSSA-N
Synonym cis-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, z-2-methyl-4-1h-purin-6-ylamino but-2-en-1-ol, z-4-7h-purin-6-yl amino-2-methylbut-2-en-1-ol, 6-4-hydroxy-3-methyl-cis-2-butenylamino purine, e-zeatin, 2z-2-methyl-4-9h-purin-6-ylamino-2-buten-1-ol, cis-zeatin hplc, z-2-methyl-4-7h-purin-6-ylamino but-2-en-1-ol
PubChem CID 688597
ChEBI CHEBI:46570
IUPAC Name (Z)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Quantity 5 mg
Formula Weight 219.2
Physical Form Solid
Chemical Name or Material cis-Zeatin
Recommended Storage Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.