Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CISD1 Antibody (1A8), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222567AF350
Description
CISD1 Monoclonal specifically detects CISD1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
CISD1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CISD1 | |
Recombinant human CISD1 (32-108aa) purified from E. coli | |
Primary | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA | |
1A8 | |
Western Blot, Flow Cytometry, ELISA | |
C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1 | |
Mouse | |
0.1 mL | |
55847 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction