Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CISD1 Antibody (1A8), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222567APCCY7
Description
CISD1 Monoclonal specifically detects CISD1 in Human samples. It is validated for Flow Cytometry.Specifications
CISD1 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CISD1 | |
Recombinant human CISD1 (32-108aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human CISD1 mAb, clone 1A8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CISD1 protein. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
1A8 | |
Flow Cytometry | |
C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1 | |
Mouse | |
Protein A purified | |
Core ESC Like Genes, Stem Cell Markers | |
55847.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction