Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CISD1 Antibody (OTI4G3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272026FR
Description
CISD1 Monoclonal specifically detects CISD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CISD1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CISD1 | |
Full length recombinant protein of human CISD1 (NP_060934) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4G3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
55847.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction