Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CISD1 Monoclonal Antibody (OTI4G3), TrueMAB™, OriGene
SDP

Catalog No. 502542681
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2681 30 μL
50-167-2068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2681 Supplier OriGene Supplier No. TA500927S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CISD1 Monoclonal antibody specifically detects CISD1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CISD1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G3
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PSMC3
Gene Accession No. P17980
Gene Alias AU043990; AW743335; C10orf70; CDGSH iron sulfur domain 1; CDGSH iron sulfur domain-containing protein 1; CDGSH iron sulfur domain-containing protein-like protein; CDGSH iron-sulfur domain-containing protein 1; Cisd1; D10Ertd214e; hypothetical protein LOC393577; hypothetical protein LOC531444; MDS029; MGC14684; MitoNEET; RGD1309529; si:dkey-18n13.4; ZCD1; zgc:63561; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1
Host Species Mouse
Immunogen Full length human recombinant protein of human PSMC3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5702
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.