Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CISD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587804 Supplier Invitrogen™ Supplier No. PA587804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAYKRFYVKD HRNKAMINLH IQKDNPKIVH AFDMEDLGDK AVYCRCWRSK KFPFCDGAHT KHNEETGDNV GPLIIKKKET; Positive Samples: HeLa, Mouse liver, Mouse heart, Mouse skeletal muscle, Rat brain; Cellular Location: Mitochondrion outer membrane, Single-pass type III membrane protein.

This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CISD1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene cisd1
Gene Accession No. B0K020, Q91WS0
Gene Alias AU043990; AW743335; C10orf70; CDGSH iron sulfur domain 1; CDGSH iron sulfur domain-containing protein 1; CDGSH iron sulfur domain-containing protein-like protein; CDGSH iron-sulfur domain-containing protein 1; Cisd1; D10Ertd214e; hypothetical protein LOC393577; hypothetical protein LOC531444; MDS029; MGC14684; MitoNEET; RGD1309529; si:dkey-18n13.4; ZCD1; zgc:63561; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1
Gene Symbols cisd1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-108 of human CISD1 (NP_0609341).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294362, 52637
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.