Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CISD2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297235PE
Description
CISD2 Polyclonal antibody specifically detects CISD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CISD2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CDGSH iron sulfur domain 2, CDGSH iron-sulfur domain-containing protein 2, CDGSH2, Endoplasmic reticulum intermembrane small protein, ERISWFS2, Miner1Wolfram syndrome 2, mitoNEET related 1, MitoNEET-related 1 protein, NAF-1, Nutrient-deprivation autophagy factor-1, ZCD2CDGSH iron sulfur domain-containing protein 2, zinc finger, CDGSH-type domain 2 | |
Produced in rabbits immunized with E. coli-derived Human CISD2 fragment. | |
0.1 mL | |
Neuroscience, Signal Transduction | |
493856 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction