Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CISD2 Mouse anti-Human, Mouse, Rat, Zebrafish, Clone: 3D7A3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CISD2 |
---|---|
Clone | 3D7A3 |
Concentration | 1.65 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6519
![]() ![]() |
Proteintech Group Inc
660821IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This monoclonal antibody is specific to CISD2 and does not cross-react with CISD1.
Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype.Specifications
CISD2 | |
1.65 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDGSH iron sulfur domain 2, CDGSH2, CISD2, CISD2-Specific, ERIS, Miner1, MitoNEET related 1 protein, NAF 1, WFS2, ZCD2 | |
cisd2 | |
Primary | |
-20°C | |
CISD2 |
3D7A3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Zebrafish | |
Q7T326, Q8N5K1, Q9CQB5 | |
295457, 393354, 493856, 67006 | |
IgG2b | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title