Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ CISD2 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531955
Description
Recommended positive controls: Drosophila melanogaster (adult wild type), Cisd2 over express (actin promoter). Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype. 
            Specifications
| CISD2 | |
| Polyclonal | |
| Unconjugated | |
| CISD2 | |
| 1500009M05Rik; 1500026J14Rik; 1500031D15Rik; AI848398; B630006A20Rik; BcDNA:RE49709; CDGSH iron sulfur domain 2; CDGSH iron sulfur domain-containing protein 2; CDGSH iron-sulfur domain-containing protein 2; CDGSH iron-sulfur domain-containing protein 2 homolog; Cdgsh2; CG1458; CG1458-PA; cisd2; Cisd2-PA; dkey-162b23.1; Dmel\CG1458; Dmel_CG1458; dWFS2; endoplasmic reticulum intermembrane small protein; ERIS; Miner1; MitoNEET; mitoNEET related 1; mitoNEET-related 1 protein; NAF-1; nervous system overexpressed protein 70; nervous system over-expressed protein 70; Noxp70; Nutrient-deprivation autophagy factor-1; RE49709p; RGD1566242; RH38554p; WFS2; Zcd2; zgc:64148; zgc:64148 protein; zinc finger, CDGSH-type domain 2; zKM0001 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 43459, 493856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N5K1, Q9VAM6 | |
| CISD2 | |
| Full length Drosophila CG1458 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Drosophila, Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                            