Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CISD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106299 Supplier Invitrogen™ Supplier No. PA5106299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CISD2.

Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CISD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CISD2
Gene Accession No. Q8N5K1, Q9CQB5
Gene Alias 1500009M05Rik; 1500026J14Rik; 1500031D15Rik; AI848398; B630006A20Rik; BcDNA:RE49709; CDGSH iron sulfur domain 2; CDGSH iron sulfur domain-containing protein 2; CDGSH iron-sulfur domain-containing protein 2; CDGSH iron-sulfur domain-containing protein 2 homolog; Cdgsh2; CG1458; CG1458-PA; cisd2; Cisd2-PA; dkey-162b23.1; Dmel\CG1458; Dmel_CG1458; dWFS2; endoplasmic reticulum intermembrane small protein; ERIS; Miner1; MitoNEET; mitoNEET related 1; mitoNEET-related 1 protein; NAF-1; nervous system overexpressed protein 70; nervous system over-expressed protein 70; Noxp70; Nutrient-deprivation autophagy factor-1; RE49709p; RGD1566242; RH38554p; WFS2; Zcd2; zgc:64148; zgc:64148 protein; zinc finger, CDGSH-type domain 2; zKM0001
Gene Symbols CISD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CISD2(Accession Q8N5K1), corresponding to amino acid residues V83-K133.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295457, 493856, 67006
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.