Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cisplatin Modified DNA Antibody (CP9/19) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$196.10
Specifications
Antigen | Cisplatin Modified DNA |
---|---|
Clone | CP9/19 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot 1:100 - 1:2000 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot BlotSpecifications
Cisplatin Modified DNA | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
DNA Repair, Epigenetics | |
PBS | |
Cisplatin modified native DNA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CP9/19 | |
Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Non-species specific | |
ICR4 | |
IgG2a | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title