Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cisplatin, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601709 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle

C1668, 15663-27-1, Cl2H6N2Pt

Spectrum™ Chemical Cisplatin, USP is part of the class of drugs known as platinum-containing anti-cancer drugs. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl

Specifications

CAS Max % 100%
Quantity 1 g
UN Number UN3288
Percent Purity 98 to 102%
Grade USP
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.