Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cit-Histone H3 (Arg2, Arg8, Arg17) Polyclonal Antibody, Abbomax

Catalog No. 502204560
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4560 Supplier Abbomax Inc Supplier No. 630180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: 3T3 Nuclear Extract Cellular location: Nuclear.

Cit-Histone H3 (Arg2, Arg8, Arg17) Polyclonal antibody specifically detects Cit-Histone H3 (Arg2, Arg8, Arg17) in Bovine, Chicken, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen Cit-Histone H3 (Arg2, Arg8, Arg17)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene H3C1
Gene Accession No. P68431, P68433, P84228
Gene Alias Citrullinated Histone H3; H3 citrulline (R2/8/17); histone H3 with citrulline modification at 2/8/17
Host Species Rabbit
Immunogen A synthetic peptide containing N-terminus of human histone H3 with citrulline modification at 2/8/17
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260423, 319148, 319149, 319150, 319152, 360198, 788250, 8350, 8351, 8352, 8355, 8357, 8358, 8968, 97908
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.