Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553919 Supplier Invitrogen™ Supplier No. PA553919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKVPRNNKRG QQGWDRKYIV LEGSKVLIYD NEAREAGQRP VEEFELCLPD GDVSIHGAVG ASELANTAKA DVPYILKMES HPHTTCWPGR TLYLL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CIT
Gene Accession No. O14578
Gene Alias C030025P15Rik; CIT; Cit-k; citron; citron (rho-interacting, serine/threonine kinase 21); citron kinase; citron Rho-interacting kinase; citron rho-interacting serine/threonine kinase; citron-K kinase; citron-N; CRIK; CRIK-SK; CTRO; KIAA0949; postsynaptic density protein (citron); rho-interacting, serine/threonine-protein kinase 21; serine/threonine kinase 21; serine/threonine-protein kinase 21; STK21
Gene Symbols CIT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CIT. Recombinant protein control fragment (Product #RP-92290).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.