Learn More
CITED1 Mouse anti-Human, Clone: OTI1C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807035

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.Specifications
CITED1 | |
Monoclonal | |
Unconjugated | |
CITED1 | |
MSG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4435 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99966 | |
CITED1 | |
Full length human recombit protein of human CITED1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.