Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CITED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565541 Supplier Invitrogen™ Supplier No. PA565541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPTTSRPALDV KGGTSPAKED ANQEMSSVAY SNLAVKDRKA VAILHYPGVA SNGTKASGAP TSSSGSPIGS PTTTPPTKP Highest antigen sequence identity to the following orthologs - mouse 77%, rat 76%.

This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CITED1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CITED1
Gene Accession No. Q99966
Gene Alias AI316840; AU019144; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1; cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; Cited1; CITED1 protein; melanocyte-specific gene 1; melanocyte-specific gene 1 protein; melanocyte-specific protein 1; melanocyte-specific protein 1; p35srj; CBP/P300-interacting transactivator 1; CITED1; Msg1; MSG1 protein; transactivator, with Glu/Asp-rich carboxy-terminal domain, 1
Gene Symbols CITED1
Host Species Rabbit
Immunogen Recombinant Human CITED1. Recombinant protein control fragment (Product #RP-106216).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.