Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985065
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5065 Supplier Bioss Supplier No. BS2990R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.

Specifications

Antigen CITED1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CITED1
Gene Accession No. Q99966
Gene Alias AI316840; AU019144; BOS_25819; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; CITED1; LOC100356232; melanocyte-specific gene 1; melanocyte-specific gene 1 protein; Melanocyte-specific protein 1; MSG1; transactivator, with Glu/Asp-rich carboxy-terminal domain, 1
Gene Symbols CITED1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CITED1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4435
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.