Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CITED2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144947 Supplier Invitrogen™ Supplier No. PA5144947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFNALMGEHI HYGAGNMNAT SGIRHAMGPG TVNGGHPPSA LAPAARFNNS QFMGPPVASQ GGSLPASMQL QKLNNQYFNH HPYPHNHYMP DLHPAAGHQM NGTNQHFRDC NPKH Antigen sequence identity: Mouse 99%; Rat 100%.

The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CITED2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CITED2
Gene Accession No. Q99967
Gene Alias AI835299; ASD8; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2; cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; Cited2; ER154-like; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; Mrg1; MRG-1; MSG1-related gene 1; Msg2; MSG-related protein 1; P35srj; VSD2
Gene Symbols CITED2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 47-160. Recombinant protein control fragment (Product #RP-109779).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10370
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.