Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED4 Antibody (HT13-2D6.3), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen CITED4
Clone HT13-2D6.3
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11041572T
SDP
View Documents
Novus Biologicals
NB11041572SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11041572
SDP
View Documents
Novus Biologicals
NB11041572
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CITED4 Monoclonal specifically detects CITED4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

CITED4
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Q96RK1
163732
N-terminal fusion protein to full-length human CITED4 [UniProt# Q96RK1]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
HT13-2D6.3
Western Blot 1:100-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature
Monoclonal
Purified
RUO
Human
cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4
CITED4
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.