Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CITED4 Antibody (HT13-2D6.3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11041572AF647
Description
CITED4 Monoclonal specifically detects CITED4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CITED4 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HT13-2D6.3 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q96RK1 | |
CITED4 | |
N-terminal fusion protein to full-length human CITED4 [UniProt# Q96RK1] | |
0.1 mL | |
Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
163732 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction