Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CITED4 Monoclonal Antibody (HT13-2D6.3), HRP
GREENER_CHOICE

Catalog No. PIMA516077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516077 Supplier Invitrogen™ Supplier No. MA516077
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: ECV304 cell lysate, transfected whole cell Hep3B lysate.

Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CITED4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HT13-2D6.3
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene CITED4
Gene Accession No. Q96RK1
Gene Alias Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4
Gene Symbols CITED4
Host Species Mouse
Immunogen N-terminal fusion protein to full-length human CITED4.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163732
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.