Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CITED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115021 Supplier Invitrogen™ Supplier No. PA5115021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CITED4.

Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CITED4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CITED4
Gene Accession No. Q96RK1, Q99MA0, Q9WUL8
Gene Alias Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4
Gene Symbols CITED4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CITED4(Accession Q96RK1), corresponding to amino acid residues S19-P69.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114491, 163732, 56222
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.