Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrate Buffer, 0.2 M Buffer Solution, pH 4.0, Low Endotoxin, PROMO, Thermo Scientific Chemicals

Catalog No. J67435AEPM
Click to view available options
Quantity:
100 mL
250 mL

Specifications

Chemical Name or Material Citrate Buffer
CAS 6132-04-3
Color Colorless
Concentration 0.2 M
pH 4.0
Physical Form Liquid
Quantity 100 mL
Molecular Formula C6H9Na3O9
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 294.1 g/mol
PubChem CID 71474
ChEBI CHEBI:32142
Show More Show Less

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.