Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Citrate Buffer Solutions
SDP

Catalog No. NB90066733 Shop All Novus Products
Change view
Click to view available options
Chemical Name or Material:
10X Antigen Retrieval Buffer High
10X Citrate Buffer
Citrate Buffer
pH:
6
7
8
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material pH
NB90066733 10X Antigen Retrieval Buffer High 10
NB90062075 10X Citrate Buffer 6
NB90066728 Citrate Buffer 7
NB90066729 10X Citrate Buffer 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB90066733 Supplier R&D Systems™ Supplier No. NB90066733

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For recovery of antigens masked by fixation in cross-linking fixatives, such as formalin

  • 10X Citrate buffer for Immunohistochemistry
  • Dilute one part buffer with nine parts distilled water
  • Unit size: 500mL

Recommended Storage

Store at room temperature

Specifications

Chemical Name or Material 10X Antigen Retrieval Buffer High
For Use With (Application) The Antigen Retrieval Protocol is Recommended for Use in Tissues That Have Been Fixed in Formalin Only. Ensure That The Fixed Sections Are Adequately Embedded in Paraffin. Cut Tissue Sections to 4-5 Microns. Preparation of Working Solutions 1. The 10 x Concentrated Format Should be Diluted Tenfold with Distilled or Deionized Water. 2. Mix One Part of Concentrated Antigen Retrieval Solution with Nine Parts of Deionized or Distilled Water. 3. Shake The Bottle Vigorously to Completely Mix The Components of The Concentrate (The Solution May Separate Into Phases Over Time). 4. Store with Cap Tightly Secured. Protocol Recommendations 1. Deparaffinize and Rehydrate Tissue Sections. 2. Place Slides Into 1 x Retrieval Solution in a Slide Container (e.g. Coplin Jar, Tissue-Tek, Staining Dish or Metal Slide Canister). 3. Retrieve Sections Under Pressure. 4. After Take-Off Reagent Jar Containing Slides From Pressure Cooker, Allow The Slides to Cool for 20 Minutes to Reach Room Temperature. 5. Wash Slides in Deionized Water and Then with Wash Buffer. Proceed with Immunostaining Recommendations in The Antibody Datasheet. 6. Gently Rinse by Gradually Adding DI Water to The Solution, Then Remove Slides and Rinse with DI Water.
pH 10
Product Line Novus
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.